Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2',4'-Dihydroxypropiophenone, 99%

Click to view available options
Quantity:
25g
100g
5792-36-9
C9H10O3
166.176
MFCD00002280
LLBBBYLDTDJMNU-UHFFFAOYSA-N
2',4'-dihydroxypropiophenone,1-2,4-dihydroxyphenyl propan-1-one,2,4-dihydroxypropiophenone,4-propionylresorcinol,1-propanone, 1-2,4-dihydroxyphenyl,respropiophenone,resorcinol fragment, 9,acmc-209m1c,2',4'-dihydroxy-propiophenone,2/',4/'-dihydroxypropiophenone
79856
1-(2,4-dihydroxyphenyl)propan-1-one
CCC(=O)C1=C(C=C(C=C1)O)O
5792-36-9
C9H10O3
166.176
MFCD00002280
LLBBBYLDTDJMNU-UHFFFAOYSA-N
2',4'-dihydroxypropiophenone,1-2,4-dihydroxyphenyl propan-1-one,2,4-dihydroxypropiophenone,4-propionylresorcinol,1-propanone, 1-2,4-dihydroxyphenyl,respropiophenone,resorcinol fragment, 9,acmc-209m1c,2',4'-dihydroxy-propiophenone,2/',4/'-dihydroxypropiophenone
79856
1-(2,4-dihydroxyphenyl)propan-1-one
CCC(=O)C1=C(C=C(C=C1)O)O
Melting Point 98°C to 101°C
Quantity 25g
Beilstein 1238635
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2',4'-Dihydroxypropiophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning