Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
626-03-9
C5H5NO2
111.1
MFCD00006273
ZEZJPIDPVXJEME-UHFFFAOYSA-N
2,4-dihydroxypyridine,pyridine-2,4-diol,2,4-pyridinediol,4-hydroxypyridin-2 1h-one,3-deazauracil,4-hydroxy-2-pyridone,2 1h-pyridinone, 4-hydroxy,2,4-dihydroxy pyridine,2-hydroxy-1h-pyridin-4-one,4-hydroxy-2 1h-pyridinone
54696004
4-hydroxy-1H-pyridin-2-one
C1=CNC(=O)C=C1O
626-03-9
C5H5NO2
111.1
MFCD00006273
ZEZJPIDPVXJEME-UHFFFAOYSA-N
2,4-dihydroxypyridine,pyridine-2,4-diol,2,4-pyridinediol,4-hydroxypyridin-2 1h-one,3-deazauracil,4-hydroxy-2-pyridone,2 1h-pyridinone, 4-hydroxy,2,4-dihydroxy pyridine,2-hydroxy-1h-pyridin-4-one,4-hydroxy-2 1h-pyridinone
54696004
4-hydroxy-1H-pyridin-2-one
C1=CNC(=O)C=C1O
Packaging Glass bottle
Color Beige-Yellow to White
Infrared Spectrum Authentic
Melting Point 278.0°C
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C5H3N)(OH)2
Beilstein 21, 160
Formula Weight 111.1
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 4-Dihydroxypyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-924-8