Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxy-1-nitrobenzene, 98%, Acros Organics

Click to view available options
Quantity:
5g
4920-84-7
C8H9NO4
183.163
MFCD00024210
XXWIYOBCHKCWNT-UHFFFAOYSA-N
1,3-dimethoxy-4-nitrobenzene,2,4-dimethoxynitrobenzene,2,4-dimethoxy-1-nitro-benzene,benzene, 2,4-dimethoxy-1-nitro,2,4-dimethoxylnitrobenzene,pubchem19759,acmc-209kfc,2,4dimethoxy-nitrobenzene,2,4-dimethoxy-nitrobenzene,2.4-dimethoxy-nitrobenzene
78633
2,4-dimethoxy-1-nitrobenzene
COC1=CC(=C(C=C1)[N+](=O)[O-])OC
4920-84-7
C8H9NO4
183.163
MFCD00024210
XXWIYOBCHKCWNT-UHFFFAOYSA-N
1,3-dimethoxy-4-nitrobenzene,2,4-dimethoxynitrobenzene,2,4-dimethoxy-1-nitro-benzene,benzene, 2,4-dimethoxy-1-nitro,2,4-dimethoxylnitrobenzene,pubchem19759,acmc-209kfc,2,4dimethoxy-nitrobenzene,2,4-dimethoxy-nitrobenzene,2.4-dimethoxy-nitrobenzene
78633
2,4-dimethoxy-1-nitrobenzene
COC1=CC(=C(C=C1)[N+](=O)[O-])OC
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Melting Point 72°C to 76°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)C6H3NO
Formula Weight 183.16
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dimethoxy-1-nitrobenzene, 98%
Show More Show Less