Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dimethoxyacetophenone, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
829-20-9
C10H12O3
180.203
MFCD00008727
VQTDPCRSXHFMOL-UHFFFAOYSA-N
2',4'-dimethoxyacetophenone,1-2,4-dimethoxyphenyl ethanone,2,4-dimethoxyacetophenone,1-2,4-dimethoxyphenyl ethan-1-one,ethanone, 1-2,4-dimethoxyphenyl,acetophenone, 2',4'-dimethoxy,peonol methyl ether,1-acetyl-2,4-dimethoxybenzene,pubchem10582,acmc-209pqs
70016
1-(2,4-dimethoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OC)OC
829-20-9
C10H12O3
180.203
MFCD00008727
VQTDPCRSXHFMOL-UHFFFAOYSA-N
2',4'-dimethoxyacetophenone,1-2,4-dimethoxyphenyl ethanone,2,4-dimethoxyacetophenone,1-2,4-dimethoxyphenyl ethan-1-one,ethanone, 1-2,4-dimethoxyphenyl,acetophenone, 2',4'-dimethoxy,peonol methyl ether,1-acetyl-2,4-dimethoxybenzene,pubchem10582,acmc-209pqs
70016
1-(2,4-dimethoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OC)OC
Color White
Packaging Glass bottle
Quantity 25g
Melting Point 38°C to 42°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H3COCH3
Beilstein 08, 267
Formula Weight 180.2
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2', 4'-Dimethoxyacetophenone
Show More Show Less