Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxyaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
100g
250g
Packaging:
Glass bottle
2735-04-8
C8H11NO2
153.18
MFCD00008371
GEQNZVKIDIPGCO-UHFFFAOYSA-N
benzenamine, 2,4-dimethoxy,aniline, 2,4-dimethoxy,2,4-dimethoxyphenylamine,unii-i079354gsi,benzenamine, 2,4-dimethoxy-, hydrochloride,2,4-domethoxyaniline,2,4-dimethoxy-anilin,2, 4-dimethoxyaniline,2,4 dimethoxy aniline,2,4-dimethoxy aniline
17652
2,4-dimethoxyaniline
COC1=CC(=C(C=C1)N)OC
2735-04-8
C8H11NO2
153.18
MFCD00008371
GEQNZVKIDIPGCO-UHFFFAOYSA-N
benzenamine, 2,4-dimethoxy,aniline, 2,4-dimethoxy,2,4-dimethoxyphenylamine,unii-i079354gsi,benzenamine, 2,4-dimethoxy-, hydrochloride,2,4-domethoxyaniline,2,4-dimethoxy-anilin,2, 4-dimethoxyaniline,2,4 dimethoxy aniline,2,4-dimethoxy aniline
17652
2,4-dimethoxyaniline
COC1=CC(=C(C=C1)N)OC
Quantity 50g
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 34°C to 37°C
Color Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3NH2
Beilstein 13, 784
Solubility Information Solubility in water: 7g/L (20°C)
Formula Weight 153.18
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Dimethoxyaniline, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 220-355-7

RTECSNumber : BX4200000

TSCA : TSCA