Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
613-45-6
C9H10O3
166.18
LWRSYTXEQUUTKW-UHFFFAOYSA-N
benzaldehyde, 2,4-dimethoxy,2,4-dimethoxybenzaldehyd,2,4-dimethoxy-benzaldehyde,2,4-bis methyloxy benzaldehyde,ameba resin,pubchem8220,acmc-209mro,2,4dimethoxybenzaldehyde,argopore tm-mb-cho,2,4-dimethoxybenzaldehye
69175
2,4-dimethoxybenzaldehyde
COC1=CC(=C(C=C1)C=O)OC
613-45-6
C9H10O3
166.18
LWRSYTXEQUUTKW-UHFFFAOYSA-N
benzaldehyde, 2,4-dimethoxy,2,4-dimethoxybenzaldehyd,2,4-dimethoxy-benzaldehyde,2,4-bis methyloxy benzaldehyde,ameba resin,pubchem8220,acmc-209mro,2,4dimethoxybenzaldehyde,argopore tm-mb-cho,2,4-dimethoxybenzaldehye
69175
2,4-dimethoxybenzaldehyde
COC1=CC(=C(C=C1)C=O)OC
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Brown
Quantity 25g
Melting Point 68.0°C to 72.0°C
Boiling Point 165.0°C (10.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H3CHO
Beilstein 08,242
Solubility Information Solubility in water: insoluble
Formula Weight 166.18
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethoxybenzaldehyde
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning