Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethoxybenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
133730-34-4
C8H11BO4
181.982
MFCD01074590
SQTUYFKNCCBFRR-UHFFFAOYSA-N
2,4-dimethoxybenzeneboronic acid,2,4-dimethoxyphenyl boronic acid,2,4-dimethoxyphenyl boranediol,boronic acid, 2,4-dimethoxyphenyl,2,4-dimethoxybenzene boronic acid,boronic acid, b-2,4-dimethoxyphenyl,pubchem1823,acmc-209btd,ksc174i6j,2,4-dimethoxyphenylboronicacid
2734341
(2,4-dimethoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC)OC)(O)O
133730-34-4
C8H11BO4
181.982
MFCD01074590
SQTUYFKNCCBFRR-UHFFFAOYSA-N
2,4-dimethoxybenzeneboronic acid,2,4-dimethoxyphenyl boronic acid,2,4-dimethoxyphenyl boranediol,boronic acid, 2,4-dimethoxyphenyl,2,4-dimethoxybenzene boronic acid,boronic acid, b-2,4-dimethoxyphenyl,pubchem1823,acmc-209btd,ksc174i6j,2,4-dimethoxyphenylboronicacid
2734341
(2,4-dimethoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC)OC)(O)O
Quantity 5g
Melting Point 115°C to 120°C
Beilstein 4744213
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning