Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzoyl chloride, 98%, Acros Organics

Click to view available options
Quantity:
5g
39828-35-8
C9H9ClO3
200.618
MFCD00017527
KVIUXRJCBBXEGJ-UHFFFAOYSA-N
2,4-dimethoxybenzoylchloride,acmc-20aocn,2, 4-dimethoxybenzoyl chloride,2,4-dimethoxy benzoyl chloride,2,4-dimethoxy-benzoyl chloride,benzoyl chloride,2,4-dimethoxy,2,4-dimethoxybenzoic acid chloride,2,4-bis methyloxy benzoyl chloride,2,4-dimethoxy-1-benzenecarbonyl chloride,benzoyl chloride, 2,4-dimethoxy-9ci
2734719
2,4-dimethoxybenzoyl chloride
COC1=CC(=C(C=C1)C(=O)Cl)OC
39828-35-8
C9H9ClO3
200.618
MFCD00017527
KVIUXRJCBBXEGJ-UHFFFAOYSA-N
2,4-dimethoxybenzoylchloride,acmc-20aocn,2, 4-dimethoxybenzoyl chloride,2,4-dimethoxy benzoyl chloride,2,4-dimethoxy-benzoyl chloride,benzoyl chloride,2,4-dimethoxy,2,4-dimethoxybenzoic acid chloride,2,4-bis methyloxy benzoyl chloride,2,4-dimethoxy-1-benzenecarbonyl chloride,benzoyl chloride, 2,4-dimethoxy-9ci
2734719
2,4-dimethoxybenzoyl chloride
COC1=CC(=C(C=C1)C(=O)Cl)OC
Quantity 5g
Color Beige to Brown
Melting Point 55°C to 60°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C(O)ClC6H3(OCH3)
Formula Weight 200.62
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dimethoxybenzoyl chloride
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger