Alfa Aesar™ 2,4-Dimethoxybenzyl alcohol, 98%

Manufacturer: Alfa Aesar™ B2435814

Quantity 25g

 View more versions of this product

Catalog No. AAB2435814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7314-44-5
    Molecular Formula C9H12O3
    Formula Weight 168.19
    MDL Number MFCD00004614
    Beilstein 2091660
    Chemical Name or Material 2,4-Dimethoxybenzyl alcohol

    Specifications

    Formula Weight 168.19
    Molecular Formula C9H12O3
    Melting Point 35° to 39°C
    Boiling Point 177° to 179°C (10mmHg)
    MDL Number MFCD00004614
    CAS 7314-44-5
    Beilstein 2091660
    Quantity 25g
    Flash Point >110°C (230°F)
    Chemical Name or Material 2,4-Dimethoxybenzyl alcohol
    Assay 98%

    Description

    Applications
    2, 4-Dimethoxybenzyl alcohol is used as a medical intermediate.

    Solubility
    Soluble in alcohol. Insoluble in water.

    Notes
    Store in refrigerator. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit