Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
20781-20-8
C9H13NO2
167.21
MFCD00052393
QOWBXWFYRXSBAS-UHFFFAOYSA-N
2,4-dimethoxybenzylamine,2,4-dimethoxyphenyl methanamine,2,4-dimethoxy benzylamine,1-2,4-dimethoxyphenyl methanamine,benzenemethanamine, 2,4-dimethoxy,2,4-dimethoxybenzyl amine,2,4-dimethoxyphenyl methylamine,2, 4-dimethoxybenzylamine,pubchem7398,2,4dimethoxybenzylamine
597250
(2,4-dimethoxyphenyl)methanamine
COC1=CC(=C(C=C1)CN)OC
20781-20-8
C9H13NO2
167.21
MFCD00052393
QOWBXWFYRXSBAS-UHFFFAOYSA-N
2,4-dimethoxybenzylamine,2,4-dimethoxyphenyl methanamine,2,4-dimethoxy benzylamine,1-2,4-dimethoxyphenyl methanamine,benzenemethanamine, 2,4-dimethoxy,2,4-dimethoxybenzyl amine,2,4-dimethoxyphenyl methylamine,2, 4-dimethoxybenzylamine,pubchem7398,2,4dimethoxybenzylamine
597250
(2,4-dimethoxyphenyl)methanamine
COC1=CC(=C(C=C1)CN)OC
Flash Point >110°C
Boiling Point 140°C
Specific Gravity 1.113
Density 1.1130g/mL
Refractive Index 1.5480 to 1.55
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H3CH2NH2
Formula Weight 167.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dimethoxybenzylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger