Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
936250-17-8
C12H19BN2O4
266.11
RKHZHTLJEUBARP-UHFFFAOYSA-N
2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronic acid pinacol ester,2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronic acid, pinacol ester,2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidine,2,4-dimethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronicacidpinacolester,2,4-dimethoxyprimidine-5-boronic acid pinacol ester,2,6-dimethoxypyrimidine-5-boronic acid, pinacol ester
16218388
2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2OC)OC
936250-17-8
C12H19BN2O4
266.11
RKHZHTLJEUBARP-UHFFFAOYSA-N
2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronic acid pinacol ester,2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronic acid, pinacol ester,2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidine,2,4-dimethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2,4-dimethoxypyrimidine-5-boronicacidpinacolester,2,4-dimethoxyprimidine-5-boronic acid pinacol ester,2,6-dimethoxypyrimidine-5-boronic acid, pinacol ester
16218388
2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2OC)OC
Melting Point 104°C to 109°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 266.11
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethoxypyrimidine-5-boronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning