Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
2510-37-4
C6H7NO3
141.13
JLSFKHJNJFXGAB-UHFFFAOYSA-N
2,4-dimethyloxazole-5-carboxylic acid,2,4-dimethyl-oxazole-5-carboxylic acid,dimethyl-1,3-oxazole-5-carboxylic acid,2,4-dimethyl-1,3-oxazole-5-carboxylicacid,2,4-dimethyl-5-oxazolecarboxylic acid,5-oxazolecarboxylic acid, 2,4-dimethyl,pubchem17689,ukrorgsyn-bb bbv-156622,2,4-dimethyl-oxazole-5-carboxylicacid,saltdata: free
1132979
2,4-dimethyl-1,3-oxazole-5-carboxylic acid
CC1=C(OC(=N1)C)C(=O)O
2510-37-4
C6H7NO3
141.13
JLSFKHJNJFXGAB-UHFFFAOYSA-N
2,4-dimethyloxazole-5-carboxylic acid,2,4-dimethyl-oxazole-5-carboxylic acid,dimethyl-1,3-oxazole-5-carboxylic acid,2,4-dimethyl-1,3-oxazole-5-carboxylicacid,2,4-dimethyl-5-oxazolecarboxylic acid,5-oxazolecarboxylic acid, 2,4-dimethyl,pubchem17689,ukrorgsyn-bb bbv-156622,2,4-dimethyl-oxazole-5-carboxylicacid,saltdata: free
1132979
2,4-dimethyl-1,3-oxazole-5-carboxylic acid
CC1=C(OC(=N1)C)C(=O)O
Melting Point 229.0°C to 231.0°C
Quantity 5g
Assay Percent Range 97%
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethyl-1,3-oxazole-5-carboxylic acid

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning