Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethyl-2-pentenoic acid, predominantly trans, 99%

Click to view available options
Quantity:
2.5g
10g
66634-97-7
C7H12O2
128.171
MFCD00209505
DMHLGGQHOSTMJG-GQCTYLIASA-N
2,4-dimethyl-2-pentenoic acid,unii-ki18c46dhr,2e-2,4-dimethylpent-2-enoic acid,ki18c46dhr,2,4-dimethyl-2-pentenoic acid, 2e,e-2,4-dimethyl-2-pentenoic acid,2,4-dimethyl-2-pentenoic acid, predominantly trans,fema no. 3143,e-2,4-dimethylpent-2-enoic acid,2-pentenoic acid, 2,4-dimethyl
5362565
(E)-2,4-dimethylpent-2-enoic acid
CC(C)C=C(C)C(=O)O
66634-97-7
C7H12O2
128.171
MFCD00209505
DMHLGGQHOSTMJG-GQCTYLIASA-N
2,4-dimethyl-2-pentenoic acid,unii-ki18c46dhr,2e-2,4-dimethylpent-2-enoic acid,ki18c46dhr,2,4-dimethyl-2-pentenoic acid, 2e,e-2,4-dimethyl-2-pentenoic acid,2,4-dimethyl-2-pentenoic acid, predominantly trans,fema no. 3143,e-2,4-dimethylpent-2-enoic acid,2-pentenoic acid, 2,4-dimethyl
5362565
(E)-2,4-dimethylpent-2-enoic acid
CC(C)C=C(C)C(=O)O
Quantity 2.5g
UN Number UN3265
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-2-pentenoic acid, predominantly trans

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA