Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-3-ethylpyrrole 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
517-22-6
C8H13N
123.2
MFCD00005222
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
2,4-dimethyl-3-ethylpyrrole,kryptopyrrole,kryptopyrrol,3-ethyl-2,4-dimethylpyrrole,cryptopyrrole,2,4-dimethyl-3-ethyl-1h-pyrrole,1h-pyrrole, 3-ethyl-2,4-dimethyl,pyrrole, 3-ethyl-2,4-dimethyl,acmc-209kvh,5-20-05-00099 beilstein handbook reference
10600
3-ethyl-2,4-dimethyl-1H-pyrrole
CCC1=C(NC=C1C)C
517-22-6
C8H13N
123.2
MFCD00005222
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
2,4-dimethyl-3-ethylpyrrole,kryptopyrrole,kryptopyrrol,3-ethyl-2,4-dimethylpyrrole,cryptopyrrole,2,4-dimethyl-3-ethyl-1h-pyrrole,1h-pyrrole, 3-ethyl-2,4-dimethyl,pyrrole, 3-ethyl-2,4-dimethyl,acmc-209kvh,5-20-05-00099 beilstein handbook reference
10600
3-ethyl-2,4-dimethyl-1H-pyrrole
CCC1=C(NC=C1C)C
Boiling Point 197°C (710.0mmHg)
Flash Point 71°C
Specific Gravity 0.913
Density 0.9130g/mL
Refractive Index 1.4960 to 1.498
Color Undesignated
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Beilstein 20, I, 47
Formula Weight 123.2
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2, 4-Dimethyl-3-ethylpyrrole, 96%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 208-234-7

RTECSNumber : UX9629500

TSCA : TSCA

Recommended Storage : Product darkens on standing