Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-6-hydroxypyrimidine +99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6622-92-0
C6H8N2O
124.14
MFCD00006105
UQFHLJKWYIJISA-UHFFFAOYSA-N
2,4-dimethyl-6-hydroxypyrimidine,2,6-dimethylpyrimidin-4-ol,4-hydroxy-2,6-dimethylpyrimidine,6-hydroxy-2,4-dimethylpyrimidine,2,6-dimethyl-4-pyrimidinol,4 1h-pyrimidinone, 2,6-dimethyl,2,6-dimethyl-4-hydroxypyrimidine,2,6-dimethylpyrimidin-4 3h-one,2,4-dimethyl-6-oxypyrimidine,t6vm dnj c1 e1
81098
2,6-dimethyl-1H-pyrimidin-4-one
CC1=CC(=O)N=C(N1)C
6622-92-0
C6H8N2O
124.14
MFCD00006105
UQFHLJKWYIJISA-UHFFFAOYSA-N
2,4-dimethyl-6-hydroxypyrimidine,2,6-dimethylpyrimidin-4-ol,4-hydroxy-2,6-dimethylpyrimidine,6-hydroxy-2,4-dimethylpyrimidine,2,6-dimethyl-4-pyrimidinol,4 1h-pyrimidinone, 2,6-dimethyl,2,6-dimethyl-4-hydroxypyrimidine,2,6-dimethylpyrimidin-4 3h-one,2,4-dimethyl-6-oxypyrimidine,t6vm dnj c1 e1
81098
2,6-dimethyl-1H-pyrimidin-4-one
CC1=CC(=O)N=C(N1)C
Melting Point 198.5°C to 200°C
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol and ethanol,slightly soluble in acetone,insoluble in hydrocarbons
Formula Weight 124.14
Physical Form Crystalline Needles
Percent Purity ≥99.0%
Chemical Name or Material 2, 4-Dimethyl-6-hydroxypyrimidine, 99+%
Show More Show Less

EINECSNumber : 229-577-9

TSCA : TSCA