Alfa Aesar™ 2',4'-Dimethylacetanilide, 98%

Manufacturer: Alfa Aesar™ H5560506

Quantity 5g

 View more versions of this product

Catalog No. AAH5560506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2050-43-3
    Formula Weight 163.22
    MDL Number MFCD00026125
    Synonym N-(2,4-Dimethylphenyl)acetamide; 2',4'-Acetoxylidide
    Chemical Name or Material 2',4'-Dimethylacetanilide

    Specifications

    Melting Point 129° to 132°C
    Formula Weight 163.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00026125
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Quantity 5g
    Color White
    Physical Form Powder
    CAS 2050-43-3
    Synonym N-(2,4-Dimethylphenyl)acetamide; 2',4'-Acetoxylidide
    Health Hazard 1 Warning
    RTECS Number AN7420000
    Chemical Name or Material 2',4'-Dimethylacetanilide
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit