Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[4-(Dimethylamino)phenyl]benzimidazole, 95%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
2562-71-2
C15H15N3
237.306
MFCD00441571
ZKBBGUJBGLTNEK-UHFFFAOYSA-N
2-p-n,n-dimethylaminophenyl-1h-benzoimidazole,4-1h-benzimidazol-2-yl-n,n-dimethylaniline,4-1h-benzoimidazol-2-yl-phenyl-dimethyl-amine,n-4-1h-benzimidazol-2-yl phenyl-n,n-dimethylamine,4-benzimidazol-2-ylphenyl dimethylamine,4-1h-1,3-benzodiazol-2-yl-n,n-dimethylaniline,cambridge id 5628722,2-p-dimethylamino phenyl benzimidazole,4-2-benzimidazolyl-n,n-dimethylaniline,2-4-dimethylaminophenyl-1h-benzimidazole
242786
4-(1H-benzimidazol-2-yl)-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2
2562-71-2
C15H15N3
237.306
MFCD00441571
ZKBBGUJBGLTNEK-UHFFFAOYSA-N
2-p-n,n-dimethylaminophenyl-1h-benzoimidazole,4-1h-benzimidazol-2-yl-n,n-dimethylaniline,4-1h-benzoimidazol-2-yl-phenyl-dimethyl-amine,n-4-1h-benzimidazol-2-yl phenyl-n,n-dimethylamine,4-benzimidazol-2-ylphenyl dimethylamine,4-1h-1,3-benzodiazol-2-yl-n,n-dimethylaniline,cambridge id 5628722,2-p-dimethylamino phenyl benzimidazole,4-2-benzimidazolyl-n,n-dimethylaniline,2-4-dimethylaminophenyl-1h-benzimidazole
242786
4-(1H-benzimidazol-2-yl)-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2
Color Yellow
Quantity 250mg
Beilstein 185491
Formula Weight 237.31
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-[4-(Dimethylamino)phenyl]benzimidazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning