Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
55499-44-0
C8H11BO2
149.984
MFCD02683101
TYONHSPZXLFWKI-UHFFFAOYSA-N
2,4-dimethylphenyl boronic acid,2,4-dimethylbenzeneboronic acid,2,4-dimethyl phenyl boronic acid,2,4-dimethylphenylboronicacid,4-dimethylphenylboronic acid,boronic acid, 2,4-dimethylphenyl,4-borono-m-xylene,pubchem9565,m-xylene-4-boronic acid,acmc-1ay9d
4198739
(2,4-dimethylphenyl)boronic acid
B(C1=C(C=C(C=C1)C)C)(O)O
55499-44-0
C8H11BO2
149.984
MFCD02683101
TYONHSPZXLFWKI-UHFFFAOYSA-N
2,4-dimethylphenyl boronic acid,2,4-dimethylbenzeneboronic acid,2,4-dimethyl phenyl boronic acid,2,4-dimethylphenylboronicacid,4-dimethylphenylboronic acid,boronic acid, 2,4-dimethylphenyl,4-borono-m-xylene,pubchem9565,m-xylene-4-boronic acid,acmc-1ay9d
4198739
(2,4-dimethylphenyl)boronic acid
B(C1=C(C=C(C=C1)C)C)(O)O
Melting Point 204°C to 208°C
Quantity 5g
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning