Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
78831-87-5
C9H11Br
199.09
MFCD03844740
WGLUZJWOTTXZIC-UHFFFAOYSA-N
1-bromomethyl-2,4-dimethylbenzene,2,4-dimethylbenzyl bromide,benzene,1-bromomethyl-2,4-dimethyl,benzene, 1-bromomethyl-2,4-dimethyl,2,4-dimethylbenzylbromide,benzene, 1-bromomethyl-2,4-dimethyl-7ci, 9ci
2734604
1-(bromomethyl)-2,4-dimethylbenzene
CC1=CC(=C(C=C1)CBr)C
78831-87-5
C9H11Br
199.09
MFCD03844740
WGLUZJWOTTXZIC-UHFFFAOYSA-N
1-bromomethyl-2,4-dimethylbenzene,2,4-dimethylbenzyl bromide,benzene,1-bromomethyl-2,4-dimethyl,benzene, 1-bromomethyl-2,4-dimethyl,2,4-dimethylbenzylbromide,benzene, 1-bromomethyl-2,4-dimethyl-7ci, 9ci
2734604
1-(bromomethyl)-2,4-dimethylbenzene
CC1=CC(=C(C=C1)CBr)C
Refractive Index 1.5680 to 1.57
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 05, III, 910
Formula Weight 199.09
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethylbenzyl bromide, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning