Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethylbenzyl chloride, 96%

Click to view available options
Quantity:
5g
25g
824-55-5
C9H11Cl
154.637
MFCD00015556
BETNPSBTDMBHCZ-UHFFFAOYSA-N
2,4-dimethylbenzyl chloride,1-chloromethyl-2,4-dimethylbenzene,4-chloromethyl-m-xylene,benzene, 1-chloromethyl-2,4-dimethyl,2,4-dimethyl-1-chloromethyl benzene,1-chloromethyl-2,4-dimethyl-benzene,acmc-209tf3,timtec-bb sbb005790,labotest-bb ltbb004402
69989
1-(chloromethyl)-2,4-dimethylbenzene
CC1=CC(=C(C=C1)CCl)C
824-55-5
C9H11Cl
154.637
MFCD00015556
BETNPSBTDMBHCZ-UHFFFAOYSA-N
2,4-dimethylbenzyl chloride,1-chloromethyl-2,4-dimethylbenzene,4-chloromethyl-m-xylene,benzene, 1-chloromethyl-2,4-dimethyl,2,4-dimethyl-1-chloromethyl benzene,1-chloromethyl-2,4-dimethyl-benzene,acmc-209tf3,timtec-bb sbb005790,labotest-bb ltbb004402
69989
1-(chloromethyl)-2,4-dimethylbenzene
CC1=CC(=C(C=C1)CCl)C
Odor Pungent
Boiling Point 103°C (15mmHg)
Refractive Index 1.5386
Density 1.06
Quantity 5g
Flash Point 96°C (204°F)
UN Number UN3265
Beilstein 742366
Formula Weight 154.64
Percent Purity 96%
Chemical Name or Material 2,4-Dimethylbenzyl chloride
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-531-7

TSCA : Yes

Recommended Storage : Ambient temperatures