Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
94-98-4
C9H13N
135.21
MFCD00025575
GBSUVYGVEQDZPG-UHFFFAOYSA-N
2,4-dimethylbenzylamine,2,4-dimethylphenyl methanamine,benzenemethanamine, 2,4-dimethyl,benzylamine, 2,4-dimethyl,2,4-dimethylbenzenemethanamine,omega-amino-psi-cumene,1-2,4-dimethylphenyl methanamine,omega-amino-1,2,4-trimethylbenzene,rarechem al bw 0350,timtec-bb sbb005795
66761
(2,4-dimethylphenyl)methanamine
CC1=CC(=C(C=C1)CN)C
94-98-4
C9H13N
135.21
MFCD00025575
GBSUVYGVEQDZPG-UHFFFAOYSA-N
2,4-dimethylbenzylamine,2,4-dimethylphenyl methanamine,benzenemethanamine, 2,4-dimethyl,benzylamine, 2,4-dimethyl,2,4-dimethylbenzenemethanamine,omega-amino-psi-cumene,1-2,4-dimethylphenyl methanamine,omega-amino-1,2,4-trimethylbenzene,rarechem al bw 0350,timtec-bb sbb005795
66761
(2,4-dimethylphenyl)methanamine
CC1=CC(=C(C=C1)CN)C
Boiling Point 218°C
Refractive Index 1.5360 to 1.539
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CH2NH2
Formula Weight 135.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethylbenzylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 202-380-5