Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethylphenol, 98%

Manufacturer:  Alfa Aesar™ A1196636

Quantity 500g

 View more versions of this product

Catalog No. AAA1196636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is used as a perfuming agent in cosmetic industry. Also used in the production of high-viscosity phosphate esters, as a feedstock for hindered phenol antioxidant and specialty modified phenolic resin manufacture. Also used as disinfectant and sanitizer on hospital premises.

Solubility
Slightly soluble in water 4068 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Store under nitrogen.
Description & Specifications

Chemical Identifiers

105-67-9
122.17
UN2261
14,10082
2,4-Dimethylphenol
C8H10O
MFCD00002233
636244
2,4-Xylenol

Specifications

94°C (201°F)
21° to 23°C
105-67-9
122.17
UN2261
14,10082
2,4-Dimethylphenol
210° to 212°C
500g
C8H10O
MFCD00002233
636244
2,4-Xylenol
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID

RTECSNumber : ZE5600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit