Alfa Aesar™ 2,4-Dimethylphenol, 98%

Manufacturer: Alfa Aesar™ A1196622

Quantity 100g

 View more versions of this product

Catalog No. AAA1196622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 105-67-9
    Molecular Formula C8H10O
    Formula Weight 122.17
    MDL Number MFCD00002233
    UN Number UN2261
    Beilstein 636244
    Merck Index 14,10082
    Synonym 2,4-Xylenol
    Chemical Name or Material 2,4-Dimethylphenol

    Specifications

    Formula Weight 122.17
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00002233
    TSCA TSCA
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.
    Flash Point 94°C (201°F)
    CAS 105-67-9
    Beilstein 636244
    Merck Index 14,10082
    Synonym 2,4-Xylenol
    RTECS Number ZE5600000
    Molecular Formula C8H10O
    UN Number UN2261
    Boiling Point 210° to 212°C
    Melting Point 21° to 23°C
    Quantity 100g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
    Chemical Name or Material 2,4-Dimethylphenol
    Assay 98%

    Description

    Applications
    It is used as a perfuming agent in cosmetic industry. Also used in the production of high-viscosity phosphate esters, as a feedstock for hindered phenol antioxidant and specialty modified phenolic resin manufacture. Also used as disinfectant and sanitizer on hospital premises.

    Solubility
    Slightly soluble in water 4068 mg/L @ 25°C (est).

    Notes
    Store away from oxidizing agents. Store under nitrogen.

    GHS H Statement
    H314-H318-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit