Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
34589-46-3,109-99-9
C8H9BrMg
209.37
ATHZRUUXJPXJNP-UHFFFAOYSA-M
2,4-dimethylphenylmagnesium bromide,2,4-dimethylphenylmagnesium bromide solution,hpgwkalajlxsdh-uhfffaoysa-m,2,4-dimethylphenylmagne-sium bromide,2,4-dimethylphenyl magnesium bromide,magnesium,1,3-dimethylbenzene-6-ide,bromide,2,4-dimethylphenylmagnesium bromide, 0.5m in 2-methf,2,4-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,2,4-dimethylphenylmagnesium bromide solution, 0.5 m in thf
13637491
magnesium;1,3-dimethylbenzene-6-ide;bromide
CC1=CC=[C-]C(=C1)C.[Mg+2].[Br-]
34589-46-3,109-99-9
C8H9BrMg
209.37
ATHZRUUXJPXJNP-UHFFFAOYSA-M
2,4-dimethylphenylmagnesium bromide,2,4-dimethylphenylmagnesium bromide solution,hpgwkalajlxsdh-uhfffaoysa-m,2,4-dimethylphenylmagne-sium bromide,2,4-dimethylphenyl magnesium bromide,magnesium,1,3-dimethylbenzene-6-ide,bromide,2,4-dimethylphenylmagnesium bromide, 0.5m in 2-methf,2,4-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,2,4-dimethylphenylmagnesium bromide solution, 0.5 m in thf
13637491
magnesium;1,3-dimethylbenzene-6-ide;bromide
CC1=CC=[C-]C(=C1)C.[Mg+2].[Br-]
Packaging AcroSeal™ Glass Bottle
Density 0.9100g/mL
Specific Gravity 0.91
Flash Point −18°C
Quantity 50mL
Color Brown
CAS Min % 9.2,86.2
CAS Max % 13.8,90.8
Linear Formula (CH3)2C6H3MgBr
Solubility Information Solubility in water: reacts
Formula Weight 209.37
Physical Form Solution
Chemical Name or Material 2, 4-Dimethylphenylmagnesium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger