Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylthiazole, 98%, Thermo Scientific™

Catalog Number AC116860250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
541-58-2
C5H7NS
113.18
MFCD00014509
OBSLLHNATPQFMJ-UHFFFAOYSA-N
2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole
10934
2,4-dimethyl-1,3-thiazole
CC1=NC(C)=CS1
541-58-2
C5H7NS
113.18
MFCD00014509
OBSLLHNATPQFMJ-UHFFFAOYSA-N
2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole
10934
2,4-dimethyl-1,3-thiazole
CC1=NC(C)=CS1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 144°C to 146°C (719.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5081 to 1.5101
Quantity 25 g
Beilstein 27, IV, 981
Specific Gravity 1.05
Merck Index 11, 3249
Solubility Information Solubility in water: miscible with ice-cold water. Other solubilities: soluble in alcohol,ether
Formula Weight 113.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylthiazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-786-9
RTECSNumber XJ4375000
TSCA TSCA
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit