Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dimethylthiophenol, 95%

Click to view available options
Quantity:
5g
25g
100g
13616-82-5
C8H10S
138.228
MFCD00010019
AMNLXDDJGGTIPL-UHFFFAOYSA-N
2,4-dimethylthiophenol,2,4-dimethyl thiophenol,benzenethiol, 2,4-dimethyl,2,4-thioxylenol,2,4-xylenethiol,4-mercapto-m-xylene,2,4-dimethylbenzene-1-thiol,4-methylthiocresol,4-thio-m-xylene,m-xylene-4-thiol
83617
2,4-dimethylbenzenethiol
CC1=CC(=C(C=C1)S)C
13616-82-5
C8H10S
138.228
MFCD00010019
AMNLXDDJGGTIPL-UHFFFAOYSA-N
2,4-dimethylthiophenol,2,4-dimethyl thiophenol,benzenethiol, 2,4-dimethyl,2,4-thioxylenol,2,4-xylenethiol,4-mercapto-m-xylene,2,4-dimethylbenzene-1-thiol,4-methylthiocresol,4-thio-m-xylene,m-xylene-4-thiol
83617
2,4-dimethylbenzenethiol
CC1=CC(=C(C=C1)S)C
Odor Unpleasant
Quantity 100g
Flash Point 87°C (189°F)
Boiling Point 207°C to 208°C
UN Number UN2810
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 2,4-Dimethylthiophenol

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA