Alfa Aesar™ 2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%

Manufacturer: Alfa Aesar™ B2179009

Quantity 10g

 View more versions of this product

Catalog No. AAB2179009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 655-07-2
    Molecular Formula C7H3F3N2O5
    Formula Weight 252.11
    MDL Number MFCD00042268
    UN Number UN2927
    Chemical Name or Material 2,4-Dinitro-1-(trifluoromethoxy)benzene

    Specifications

    Formula Weight 252.11
    Quantity 10g
    Molecular Formula C7H3F3N2O5
    MDL Number MFCD00042268
    UN Number UN2927
    Health Hazard 1 Danger
    Flash Point 154°C (309°F)
    Melting Point −21°C
    CAS 655-07-2
    Health Hazard 2 GHS H Statement
    H300-H310-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
    Boiling Point 273° to 274°C
    Chemical Name or Material 2,4-Dinitro-1-(trifluoromethoxy)benzene
    Assay 98%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H300-H310-H314-H318
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit