Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%

Click to view available options
Quantity:
2.5g
10g
655-07-2
C7H3F3N2O5
252.105
MFCD00042268
KYUUZCUXYXNPFO-UHFFFAOYSA-N
2,4-dinitro-1-trifluoromethoxy benzene,2,4-dinitro trifluoromethoxy benzene,2,4-dinitrotrifluoroanisole,2,4-dinitrotrifluoromethoxybenzene,2,4-dinitro-1-trifluoromethoxy-benzene,acmc-20aoc3,2,4-dinitro trifluoroanisole,a,a,a-trifluoro-2,4-dinitro-anisole,1-trifluoromethoxy-2,4-dinitrobenzene,benzene,2,4-dinitro-1-trifluoromethoxy
2737131
2,4-dinitro-1-(trifluoromethoxy)benzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])OC(F)(F)F
655-07-2
C7H3F3N2O5
252.105
MFCD00042268
KYUUZCUXYXNPFO-UHFFFAOYSA-N
2,4-dinitro-1-trifluoromethoxy benzene,2,4-dinitro trifluoromethoxy benzene,2,4-dinitrotrifluoroanisole,2,4-dinitrotrifluoromethoxybenzene,2,4-dinitro-1-trifluoromethoxy-benzene,acmc-20aoc3,2,4-dinitro trifluoroanisole,a,a,a-trifluoro-2,4-dinitro-anisole,1-trifluoromethoxy-2,4-dinitrobenzene,benzene,2,4-dinitro-1-trifluoromethoxy
2737131
2,4-dinitro-1-(trifluoromethoxy)benzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])OC(F)(F)F
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Flash Point 154°C (309°F)
Boiling Point 273°C to 274°C
Quantity 2.5g
Melting Point -21°C
UN Number UN2927
Formula Weight 252.11
Percent Purity 98%
Chemical Name or Material 2,4-Dinitro-1-(trifluoromethoxy)benzene

GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.