Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitroaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
97-02-9
C6H5N3O4
183.12
MFCD00007151
LXQOQPGNCGEELI-UHFFFAOYSA-N
benzenamine, 2,4-dinitro,1-amino-2,4-dinitrobenzene,2,4-dinitroanilin,2,4-dinitrobenzenamine,2,4-dinitroanilina,2,4-dinitraniline,2,4-dinitrophenylamine,aniline, 2,4-dinitro,2,4-dinitroaminobenzene,2,4-nitroaniline
7321
CHEBI:34242
2,4-dinitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N
97-02-9
C6H5N3O4
183.12
MFCD00007151
LXQOQPGNCGEELI-UHFFFAOYSA-N
benzenamine, 2,4-dinitro,1-amino-2,4-dinitrobenzene,2,4-dinitroanilin,2,4-dinitrobenzenamine,2,4-dinitroanilina,2,4-dinitraniline,2,4-dinitrophenylamine,aniline, 2,4-dinitro,2,4-dinitroaminobenzene,2,4-nitroaniline
7321
CHEBI:34242
2,4-dinitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])N
Melting Point 177°C to 180°C
Flash Point 224°C
Packaging Glass bottle
Water 8 to 15% (K.F.)
Color Brown-Yellow or Green-Yellow to Yellow
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (O2N)2C6H3NH2
Beilstein 12,747
Merck Index 15,3296
Solubility Information Solubility in water: 0.06g/L (20 c). Other solubilities: 0.7% ethanol (21 c),soluble in diethyl ether
Formula Weight 183.12
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 4-Dinitroaniline, 99%
Show More Show Less

GHS H Statement:
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-553-5

RTECSNumber : BX9100000

TSCA : TSCA