Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrofluorobenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
70-34-8
C6H3FN2O4
186.1
MFCD00007056
LOTKRQAVGJMPNV-UHFFFAOYSA-N
2,4-dinitrofluorobenzene,dinitrofluorobenzene,fdnb,dnfb,2,4-dinitro-1-fluorobenzene,sanger's reagent,fluoro-2,4-dinitrobenzene,2,4-dinitrobenzenefluoride,2,4-dinitrophenyl fluoride,fluorodinitrobenzene
6264
CHEBI:53049
1-fluoro-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F
70-34-8
C6H3FN2O4
186.1
MFCD00007056
LOTKRQAVGJMPNV-UHFFFAOYSA-N
2,4-dinitrofluorobenzene,dinitrofluorobenzene,fdnb,dnfb,2,4-dinitro-1-fluorobenzene,sanger's reagent,fluoro-2,4-dinitrobenzene,2,4-dinitrobenzenefluoride,2,4-dinitrophenyl fluoride,fluorodinitrobenzene
6264
CHEBI:53049
1-fluoro-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F
Alkylation reagent
Melting Point 23°C to 26°C
Boiling Point 296°C
Flash Point >112°C
Specific Gravity 1.48
Refractive Index 1.5680 to 1.57
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
Color Brown to Yellow
Density 1.4800g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (O2N)2C6H3F
Beilstein 05,262
Fieser 01,321; 02,175; 06,233
Merck Index 15,4204
Solubility Information Solubility in water: 400mg/l (25°C). Other solubilities: soluble in ether,benzene,propylene glycol,and hot alcohol
Formula Weight 186.1
Physical Form Liquid or Low Melting Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dinitrofluorobenzene
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic if swallowed.
Suspected of causing cancer.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger