Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dinitrophenyl thiocyanate, 98%

Click to view available options
Quantity:
25g
100g
1594-56-5
C7H3N3O4S
225.178
MFCD00047702
XQDQRCRASHAZBA-UHFFFAOYSA-N
dntb,nirit,trirodazeen,gryzbol,grzybol,2,4-dinitrothiocyanobenzene,nirit supra,tri-rodazene,2,4-dinitrothiocyanatobenzene,rhodandinitrobenzol
15325
CHEBI:53051
(2,4-dinitrophenyl) thiocyanate
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N
1594-56-5
C7H3N3O4S
225.178
MFCD00047702
XQDQRCRASHAZBA-UHFFFAOYSA-N
dntb,nirit,trirodazeen,gryzbol,grzybol,2,4-dinitrothiocyanobenzene,nirit supra,tri-rodazene,2,4-dinitrothiocyanatobenzene,rhodandinitrobenzol
15325
CHEBI:53051
(2,4-dinitrophenyl) thiocyanate
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N
Quantity 100g
Odor Irritating
Melting Point 138°C to 140°C
Beilstein 1983292
Formula Weight 225.18
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrophenyl thiocyanate

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XK9450000