Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,4-Dinitrophenylhydrazine, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
100 g
500 g
10 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical 2,4-Dinitrophenylhydrazine is a chemical compound that is usually supplied as a wet red-to-orange solid that is shock sensitive. It is used in Brady′s test to detect the carbonyl functionality of an aldehyde or ketone functional group.
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 100 g |
UN Number | UN1325 |
Grade | Reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction