Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dipyridyl, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
581-47-5
C10H8N2
156.188
MFCD00006217
RMHQDKYZXJVCME-UHFFFAOYSA-N
2,4'-bipyridine,2,4'-dipyridyl,4-pyridylpyridine,2,4'-bipyridyl,2,4-bipyridyl,2,4'-dipyridine,unii-c6sn6ru851,ccris 3428,2-4-pyridyl pyridine,4-pyridyl-2-pyridine
68488
2-pyridin-4-ylpyridine
C1=CC=NC(=C1)C2=CC=NC=C2
581-47-5
C10H8N2
156.188
MFCD00006217
RMHQDKYZXJVCME-UHFFFAOYSA-N
2,4'-bipyridine,2,4'-dipyridyl,4-pyridylpyridine,2,4'-bipyridyl,2,4-bipyridyl,2,4'-dipyridine,unii-c6sn6ru851,ccris 3428,2-4-pyridyl pyridine,4-pyridyl-2-pyridine
68488
2-pyridin-4-ylpyridine
C1=CC=NC(=C1)C2=CC=NC=C2
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 58°C to 62°C
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Fieser 16,25
Formula Weight 156.19
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4'-Dipyridyl
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-467-7

TSCA : TSCA