Alfa Aesar™ 2-(4-Ethylphenyl)-5-n-propylpyrimidine, 99+%

Manufacturer: Alfa Aesar™ H2736103

Quantity 1g

 View more versions of this product

Catalog No. AAH2736103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98495-11-5
    Molecular Formula C15H18N2
    Formula Weight 226.32
    MDL Number MFCD00799380
    Synonym 5-n-Propyl-2-pyrimidinethiol
    Chemical Name or Material 2-(4-Ethylphenyl)-5-n-propylpyrimidine

    Specifications

    Formula Weight 226.32
    Molecular Formula C15H18N2
    MDL Number MFCD00799380
    Synonym 5-n-Propyl-2-pyrimidinethiol
    Melting Point 41° to 43°C
    Quantity 1g
    CAS 98495-11-5
    Chemical Name or Material 2-(4-Ethylphenyl)-5-n-propylpyrimidine
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit