Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Fluorophenoxy)nicotinic acid, 97+%

Click to view available options
Quantity:
1g
5g
54629-13-9
C12H8FNO3
233.198
MFCD00833409
SDZUYDOXBXHDCE-UHFFFAOYSA-N
2-4-fluorophenoxy nicotinic acid,2-4-fluorophenoxy pyridine-3-carboxylic acid,2-4-fluoro-phenoxy-nicotinic acid,pubchem16292,acmc-1akqk,maybridge1_000048,2-4-fluorophenoxy-3-pyridinecarboxylic acid,3-pyridinecarboxylicacid, 2-4-fluorophenoxy,3-pyridinecarboxylic acid, 2-4-fluorophenoxy
605123
2-(4-fluorophenoxy)pyridine-3-carboxylic acid
C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O
54629-13-9
C12H8FNO3
233.198
MFCD00833409
SDZUYDOXBXHDCE-UHFFFAOYSA-N
2-4-fluorophenoxy nicotinic acid,2-4-fluorophenoxy pyridine-3-carboxylic acid,2-4-fluoro-phenoxy-nicotinic acid,pubchem16292,acmc-1akqk,maybridge1_000048,2-4-fluorophenoxy-3-pyridinecarboxylic acid,3-pyridinecarboxylicacid, 2-4-fluorophenoxy,3-pyridinecarboxylic acid, 2-4-fluorophenoxy
605123
2-(4-fluorophenoxy)pyridine-3-carboxylic acid
C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O
Melting Point 182°C to 185°C
Quantity 5g
Formula Weight 233.2
Percent Purity ≥97%
Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning