Alfa Aesar™ 2,4-Heptadien-1-ol

Manufacturer: Alfa Aesar™ 1576514

Quantity 25g

 View more versions of this product

Catalog No. AA1576514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 33467-79-7
    Formula Weight 112.17
    MDL Number MFCD00014049
    Chemical Name or Material 2,4-Heptadien-1-ol

    Specifications

    Formula Weight 112.17
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Quantity 25g
    TSCA TSCA
    CAS 33467-79-7
    Health Hazard 1 Warning
    Flash Point 63° (145°F)
    MDL Number MFCD00014049
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Chemical Name or Material 2,4-Heptadien-1-ol

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit