Alfa Aesar™ 2,4-Hexadiene, tech. 90%, mixture of isomers

Manufacturer: Alfa Aesar™ H5340414

Quantity 25g

 View more versions of this product

Catalog No. AAH5340414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 592-46-1
    Molecular Formula C6H10
    Linear Formula CH3CH=CHCH=CHCH3
    Formula Weight 82.15
    MDL Number MFCD00009297
    UN Number UN2458
    Chemical Name or Material 2,4-Hexadiene, mixture of isomers
    Grade Technical Grade

    Specifications

    Formula Weight 82.15
    Linear Formula CH3CH=CHCH=CHCH3
    MDL Number MFCD00009297
    CAS 592-46-1
    UN Number UN2458
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −14°C (−7°F)
    Quantity 25g
    Molecular Formula C6H10
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXADIENES
    Boiling Point 82°C
    Chemical Name or Material 2,4-Hexadiene, mixture of isomers
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    2,4-Hexadiene is used in the production of 3,6-dimethyl-3,6-dihydro-[1,2]dithiine.

    Solubility
    Miscible with ethanol, ethyl ether and chloroform. Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit