Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Hexadienoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
500g
1kg
Packaging:
Plastic bottle
110-44-1
C6H8O2
112.13
MFCD00002703
WSWCOQWTEOXDQX-MQQKCMAXSA-N
sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid
643460
CHEBI:38358
(2E,4E)-hexa-2,4-dienoic acid
CC=CC=CC(=O)O
110-44-1
C6H8O2
112.13
MFCD00002703
WSWCOQWTEOXDQX-MQQKCMAXSA-N
sorbic acid,2,4-hexadienoic acid,2e,4e-hexa-2,4-dienoic acid,2e,4e-hexadienoic acid,panosorb,2-propenylacrylic acid,trans,trans-sorbic acid,sorbistat,hexadienoic acid,e,e-2,4-hexadienoic acid
643460
CHEBI:38358
(2E,4E)-hexa-2,4-dienoic acid
CC=CC=CC(=O)O
Flash Point 127°C
Quantity 250g
Melting Point 132°C to 135°C
Water 0.5% max. (K.F.)
Packaging Plastic bottle
Color Cream-White or White
Infrared Spectrum Authentic
Sulfated Ash 0.2% max.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH=CHCH=CHCO2H
Beilstein 02, 483
Fieser 15,296
Merck Index 15, 8847
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in ethanol,chloroform,very soluble in diethyl ether
Formula Weight 112.13
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5% (on anhydrous substance)
Chemical Name or Material 2, 4-Hexadienoic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 203-768-7

RTECSNumber : WG2100000

TSCA : TSCA