Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride, 95%

Click to view available options
Quantity:
1g
5g
25g
57591-61-4
C9H12ClNO3
217.649
MFCD01862152
UYCKVJUNDXPDJH-DDWIOCJRSA-N
r-amino-4-hydroxyphenyl acetic acid methyl ester hydrochloride,r-methyl 2-amino-2-4-hydroxyphenyl acetate hydrochloride,methyl 2r-2-amino-2-4-hydroxyphenyl acetate hydrochloride,d-4-hydroxyphenylglycine methyl ester hydrochloride,r-methyl 2-amino-2-4-hydroxyphenyl acetate hcl,d-p-hydroxyphenylglycine methyl ester hydrochloride,ksc495s1b,methyl r-amino-4-hydroxyphenyl acetate hydrochloride,methyl r-amino-4-hydroxyphenyl acetate hydrochloride,methyl 2r-amino 4-hydroxyphenyl acetate-hydrogen chloride 1/1
13215089
methyl (2R)-2-amino-2-(4-hydroxyphenyl)acetate;hydrochloride
COC(=O)C(C1=CC=C(C=C1)O)N.Cl
57591-61-4
C9H12ClNO3
217.649
MFCD01862152
UYCKVJUNDXPDJH-DDWIOCJRSA-N
r-amino-4-hydroxyphenyl acetic acid methyl ester hydrochloride,r-methyl 2-amino-2-4-hydroxyphenyl acetate hydrochloride,methyl 2r-2-amino-2-4-hydroxyphenyl acetate hydrochloride,d-4-hydroxyphenylglycine methyl ester hydrochloride,r-methyl 2-amino-2-4-hydroxyphenyl acetate hcl,d-p-hydroxyphenylglycine methyl ester hydrochloride,ksc495s1b,methyl r-amino-4-hydroxyphenyl acetate hydrochloride,methyl r-amino-4-hydroxyphenyl acetate hydrochloride,methyl 2r-amino 4-hydroxyphenyl acetate-hydrogen chloride 1/1
13215089
methyl (2R)-2-amino-2-(4-hydroxyphenyl)acetate;hydrochloride
COC(=O)C(C1=CC=C(C=C1)O)N.Cl
Formula Weight 217.65
Physical Form Solid
Percent Purity 95%
Melting Point 187°C to 190°C
Quantity 25g
Chemical Name or Material 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride