Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Hydroxyphenyl)propionic acid, 98%

Click to view available options
Quantity:
10g
50g
938-96-5
C9H10O3
166.176
MFCD00128847
ZHMMPVANGNPCBW-UHFFFAOYSA-N
2-4-hydroxyphenyl propanoic acid,2-4-hydroxyphenyl propionic acid,4-hydroxyhydratropate,4-hydroxyhydratropic acid,4-hydroxyphenyl-2-propionic acid,2-4'-hydroxyphenyl-propionic acid,2-4-hydroxyphenyl propionate,acmc-209ro0,4-hydroxyphenyl propanoic acid,methyl 4-hydroxyphenylacetic acid
102526
CHEBI:1868
2-(4-hydroxyphenyl)propanoic acid
CC(C1=CC=C(C=C1)O)C(=O)O
938-96-5
C9H10O3
166.176
MFCD00128847
ZHMMPVANGNPCBW-UHFFFAOYSA-N
2-4-hydroxyphenyl propanoic acid,2-4-hydroxyphenyl propionic acid,4-hydroxyhydratropate,4-hydroxyhydratropic acid,4-hydroxyphenyl-2-propionic acid,2-4'-hydroxyphenyl-propionic acid,2-4-hydroxyphenyl propionate,acmc-209ro0,4-hydroxyphenyl propanoic acid,methyl 4-hydroxyphenylacetic acid
102526
CHEBI:1868
2-(4-hydroxyphenyl)propanoic acid
CC(C1=CC=C(C=C1)O)C(=O)O
Quantity 10g
Melting Point 130°C to 132°C
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-(4-Hydroxyphenyl)propionic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning