Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Lutidine, 98+%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
108-47-4
C7H9N
107.15
MFCD00006337
JYYNAJVZFGKDEQ-UHFFFAOYSA-N
2,4-lutidine,pyridine, 2,4-dimethyl,alpha,gamma-dimethylpyridine,2,4-lutidene,unii-83903uj0ww,2,4-dimethyl-pyridine,.alpha.,.gamma.-dimethylpyridine,2,4-dimethyl pyridine,2.4-lutidine,2,4 lutidine
7936
2,4-dimethylpyridine
CC1=CC(=NC=C1)C
108-47-4
C7H9N
107.15
MFCD00006337
JYYNAJVZFGKDEQ-UHFFFAOYSA-N
2,4-lutidine,pyridine, 2,4-dimethyl,alpha,gamma-dimethylpyridine,2,4-lutidene,unii-83903uj0ww,2,4-dimethyl-pyridine,.alpha.,.gamma.-dimethylpyridine,2,4-dimethyl pyridine,2.4-lutidine,2,4 lutidine
7936
2,4-dimethylpyridine
CC1=CC(=NC=C1)C
Melting Point -60.0°C
Packaging Glass bottle
Density 0.9200g/mL
Boiling Point 159.0°C
Flash Point 47°C
Specific Gravity 0.92
Refractive Index 1.4990 to 1.5010
Quantity 500mL
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Beilstein , 244
Fieser 06,224
Solubility Information Solubility in water: 15g/100 g (20°C). Other solubilities: soluble in alcohol,ether,acetone and,benzene
Formula Weight 107.15
Percent Purity 98+%
Chemical Name or Material 2, 4-Lutidine, 98+%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-586-8

RTECSNumber : OK9400000

TSCA : TSCA