Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxybenzoyl)thiophene 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
4160-63-8
C12H10O2S
218.27
MFCD00005440
KYVBFEMQEUXVQB-UHFFFAOYSA-N
2-4-methoxybenzoyl thiophene,4-methoxyphenyl thiophen-2-yl methanone,2-p-methoxybenzoyl thiophene,p-anisyl thiophen-2-yl ketone,4-methoxyphenyl 2-thienyl ketone,methanone, 4-methoxyphenyl-2-thienyl,4-methoxyphenyl 2-thienyl methanone,pubchem10833,acmc-1aikh,2-thienyl p-anisyl ketone
77812
(4-methoxyphenyl)-thiophen-2-ylmethanone
COC1=CC=C(C=C1)C(=O)C2=CC=CS2
4160-63-8
C12H10O2S
218.27
MFCD00005440
KYVBFEMQEUXVQB-UHFFFAOYSA-N
2-4-methoxybenzoyl thiophene,4-methoxyphenyl thiophen-2-yl methanone,2-p-methoxybenzoyl thiophene,p-anisyl thiophen-2-yl ketone,4-methoxyphenyl 2-thienyl ketone,methanone, 4-methoxyphenyl-2-thienyl,4-methoxyphenyl 2-thienyl methanone,pubchem10833,acmc-1aikh,2-thienyl p-anisyl ketone
77812
(4-methoxyphenyl)-thiophen-2-ylmethanone
COC1=CC=C(C=C1)C(=O)C2=CC=CS2
Color Undesignated
Quantity 25g
Melting Point 74°C
Formula Weight 218.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-(4-Methoxybenzoyl)thiophene