Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Methoxyphenyl)acetamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
1055926-31-2
C9H12N2O2
180.207
QIACLRIKXICINP-UHFFFAOYSA-N
n'-hydroxy-2-4-methoxyphenyl ethanimidamide,1z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide,n-hydroxy-2-4-methoxy-phenyl-acetamidine,z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide,2-4-methoxyphenyl-acetamidoxime,benzeneethanimidamide, n-hydroxy-4-methoxy,z-n'-hydroxy-2-4-methoxyphenyl acetimidamide,benzeneethanimidamide,n-hydroxy-4-methoxy,1z-n\'-hydroxy-2-4-methoxyphenyl ethanimidamide
5356867
N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
COC1=CC=C(C=C1)CC(=NO)N
1055926-31-2
C9H12N2O2
180.207
QIACLRIKXICINP-UHFFFAOYSA-N
n'-hydroxy-2-4-methoxyphenyl ethanimidamide,1z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide,n-hydroxy-2-4-methoxy-phenyl-acetamidine,z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide,2-4-methoxyphenyl-acetamidoxime,benzeneethanimidamide, n-hydroxy-4-methoxy,z-n'-hydroxy-2-4-methoxyphenyl acetimidamide,benzeneethanimidamide,n-hydroxy-4-methoxy,1z-n\'-hydroxy-2-4-methoxyphenyl ethanimidamide
5356867
N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
COC1=CC=C(C=C1)CC(=NO)N
Color White
Quantity 250mg
Formula Weight 180.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxyphenyl)acetamidoxime