Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Methylphenyl)acetamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
1312766-67-8
C9H12N2O
164.208
CKMSCSXWLFGEGO-UHFFFAOYSA-N
n-hydroxy-2-p-tolyl-acetamidine,2-p-tolyl-acetamidoxime,2-4-tolyl-acetamidoxime,z-n'-hydroxy-2-p-tolyl acetimidamide
9679732
N'-hydroxy-2-(4-methylphenyl)ethanimidamide
CC1=CC=C(C=C1)CC(=NO)N
1312766-67-8
C9H12N2O
164.208
CKMSCSXWLFGEGO-UHFFFAOYSA-N
n-hydroxy-2-p-tolyl-acetamidine,2-p-tolyl-acetamidoxime,2-4-tolyl-acetamidoxime,z-n'-hydroxy-2-p-tolyl acetimidamide
9679732
N'-hydroxy-2-(4-methylphenyl)ethanimidamide
CC1=CC=C(C=C1)CC(=NO)N
Color Cream or White
Quantity 1g
Formula Weight 164.2
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(4-Methylphenyl)acetamidoxime