Alfa Aesar™ 2-(4-Morpholinyl)pyridine, 96%

Manufacturer: Alfa Aesar™ H5598903

Quantity 1g

 View more versions of this product

Catalog No. AAH5598903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24255-25-2
    Formula Weight 164.21
    MDL Number MFCD01143321
    Synonym 2-Morpholinopyridine
    Chemical Name or Material 2-(4-Morpholinyl)pyridine

    Specifications

    Formula Weight 164.21
    Density 1.131g/mL
    Physical Form Liquid
    MDL Number MFCD01143321
    Boiling Point 135°C (20mmHg)
    Quantity 1g
    CAS 24255-25-2
    Synonym 2-Morpholinopyridine
    Chemical Name or Material 2-(4-Morpholinyl)pyridine
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit