Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
480424-77-9,109-99-9
C11H14BrMgNO
280.44
NWGRMLFTJZVEAO-UHFFFAOYSA-M
2-4-morpholinylmethyl phenyl magnesium bromide solution,rsbwikctzydpjq-uhfffaoysa-m,2-4-morpholinylmethyl phenyl magnesiu,2-4-morpholinylmethyl phenyl magnesiu&,2-4-morpholinylmethyl phenyl magnesium bromide,2-4-morpholino methyl phenylmagnesium bromide,4-2-bromomagnesio phenyl methyl morpholine,2-4-morpholinylmethyl phenyl magnesium bromide solution, 0.25 m in thf,2-4-morpholino methyl phenylmagnesium bromide 0.25 m in tetrahydrofuran,2-4-morpholino methyl phenylmagnesium bromide, 0.25m in tetrahydrofuran
16217254
magnesium;4-(phenylmethyl)morpholine;bromide
C1COCCN1CC2=CC=CC=[C-]2.[Mg+2].[Br-]
480424-77-9,109-99-9
C11H14BrMgNO
280.44
NWGRMLFTJZVEAO-UHFFFAOYSA-M
2-4-morpholinylmethyl phenyl magnesium bromide solution,rsbwikctzydpjq-uhfffaoysa-m,2-4-morpholinylmethyl phenyl magnesiu,2-4-morpholinylmethyl phenyl magnesiu&,2-4-morpholinylmethyl phenyl magnesium bromide,2-4-morpholino methyl phenylmagnesium bromide,4-2-bromomagnesio phenyl methyl morpholine,2-4-morpholinylmethyl phenyl magnesium bromide solution, 0.25 m in thf,2-4-morpholino methyl phenylmagnesium bromide 0.25 m in tetrahydrofuran,2-4-morpholino methyl phenylmagnesium bromide, 0.25m in tetrahydrofuran
16217254
magnesium;4-(phenylmethyl)morpholine;bromide
C1COCCN1CC2=CC=CC=[C-]2.[Mg+2].[Br-]
CAS Min % 4.54,89.4
CAS Max % 10.6,95.46
Solubility Information Solubility in water: reacts
Formula Weight 280.44
Physical Form Solution
Concentration 0.15 to 0.35M
Density 0.9260g/mL
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.926
Color Yellow
Quantity 50mL
Flash Point −17°C
Chemical Name or Material [2-(4-Morpholinylmethyl)phenyl]magnesium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger