Alfa Aesar™ 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine, 97%

Manufacturer: Alfa Aesar™ B2260103

Quantity 1g

 View more versions of this product

Catalog No. AAB2260103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57202-48-9
    Molecular Formula C24H36N2O
    Formula Weight 368.56
    MDL Number MFCD00691439
    Chemical Name or Material 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine

    Specifications

    Formula Weight 368.56
    Molecular Formula C24H36N2O
    MDL Number MFCD00691439
    CAS 57202-48-9
    Melting Point 57° to 62°C
    Quantity 1g
    Chemical Name or Material 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit