Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(4-Nitrophenyl)ethanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is a phosphate protecting reagent and O6 of deoxyguanosine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
It is a phosphate protecting reagent and O6 of deoxyguanosine.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
Specifications
Melting Point | 59°C to 62°C |
Boiling Point | 177°C (16 mmHg) |
Quantity | 5 g |
Beilstein | 1866148 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 167.16 |
Percent Purity | 98% |
Chemical Name or Material | 2-(4-Nitrophenyl)ethanol |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H301-H319 Toxic if swallowed. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P280-P301+P310-P305+P351+P338-P321-P405-P501a Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment (see label). Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 202-835-8 |
RTECSNumber | SG8602000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction