Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(4-Nitrophenyl)ethylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
Solubility
Soluble in methanol almost transparency.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.
Solubility
Soluble in methanol almost transparency.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 200°C (literature) |
Quantity | 5 g |
Beilstein | 3568915 |
Solubility Information | Soluble in methanol almost transparency. |
Formula Weight | 202.64 |
Percent Purity | ≥98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2-(4-Nitrophenyl)ethylamine hydrochloride |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 249-980-3 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction