Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Nitrophenyl)ethylamine hydrochloride, ≥98%

Click to view available options
Quantity:
1g
5g
25g
29968-78-3
C8H11ClN2O2
202.638
MFCD00012900
JVMHULJEYUQYSH-UHFFFAOYSA-N
4-nitrophenethylamine hydrochloride,4-nitrophenylethylamine hydrochloride,4-nitrophenethylamine hcl,2-4-nitrophenyl ethanamine hydrochloride,2-4-nitrophenyl ethylamine hydrochloride,p-nitrophenylethylamine hcl,benzeneethanamine, 4-nitro-, monohydrochloride,4-nitrophenethylaminehydrochloride,p-nitrophenethylamine, hydrochloride,pubchem11627
458983
2-(4-nitrophenyl)ethanamine;hydrochloride
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
29968-78-3
C8H11ClN2O2
202.638
MFCD00012900
JVMHULJEYUQYSH-UHFFFAOYSA-N
4-nitrophenethylamine hydrochloride,4-nitrophenylethylamine hydrochloride,4-nitrophenethylamine hcl,2-4-nitrophenyl ethanamine hydrochloride,2-4-nitrophenyl ethylamine hydrochloride,p-nitrophenylethylamine hcl,benzeneethanamine, 4-nitro-, monohydrochloride,4-nitrophenethylaminehydrochloride,p-nitrophenethylamine, hydrochloride,pubchem11627
458983
2-(4-nitrophenyl)ethanamine;hydrochloride
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color Undesignated
Quantity 25g
Melting Point 200°C (literature)
Beilstein 3568915
Formula Weight 202.64
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 2-(4-Nitrophenyl)ethylamine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning